Imprimer
Cryogenic tubes, TruCool™
Fournisseur: BIOCISION
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BCS-2510 BCS-2511 BCS-2512 BCS-2513 BCS-2514 BCS-2515 BCS-2516 BCS-2517 BCS-2502 BCS-2501 BCS-2503 BCS-2504 BCS-2505
479-1910EA 753 EUR
479-1910 479-1911 479-1912 479-1913 479-1914 479-1915 479-1916 479-1917 479-1906 479-1905 479-1907 479-1908 479-1909
Cryogenic tubes, TruCool™
Fioles
TruCool® leak-proof cryogenic vials feature a screw cap with a thermally-fused gasket to ensure a tight seal. The gasket is a co-molded thermoplastic elastomer (TPE) layer, which prevents leaking, slipping and risk of contamination. The gasket is 95 kPa certified to provide a leak-proof seal. These vials are sterile and individually barcoded with a unique identifier which can be read with common barcode readers. Ideal for use in CoolRack® CF cryogenic vial modules and CoolCell® alcohol-free cell freezing containers. Recommended for use in vapor phase LN₂ only.
Cdt: Cases contain 5 packs of 100 vials.