Imprimer
Adapters, PTFE, socket on cone
Fournisseur: COWIE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
021.01014 021.01019 021.01419 021.01424 021.01924 021.01929 021.01934 021.02429 021.02434 021.02934 021.2445 021.2945
COWI021.01014EA 18.9 EUR
COWI021.01014 COWI021.01019 COWI021.01419 COWI021.01424 COWI021.01924 COWI021.01929 COWI021.01934 COWI021.02429 COWI021.02434 COWI021.02934 COWI021.2445 COWI021.2945
Adapters, PTFE, socket on cone
Adaptateurs Raccords à rodage conique
Made of pure PTFE, white.
  • Very good chemical resistance
  • Very good temperature resistance
  • Fully adjustable with replacement Viton® seal
  • Contact materials are glass and PTFE only
This holder enables standard probes and thermometers to be assembled in standard taper laboratory glassware.