Imprimer
Extraction thimbles
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05405225 05405243 05405249
LENZ05405225EA 58.6 EUR
LENZ05405225 LENZ05405243 LENZ05405249
Extraction thimbles
Tubes Tubes à essais
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Glass tubes with sintered filter disc (porosity: 0), for extractors acc. to Thielepape.