Imprimer
Extractor heads for specific light solvents
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05432137 05432149 05432158
LENZ05432137EA 319 EUR
LENZ05432137 LENZ05432149 LENZ05432158
Extractor heads for specific light solvents
Extracteurs Extracteurs liquide-liquide
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of liquids with specific light solvents, with insert filter (porosity: 0).