Imprimer
Flasks, round bottom, with three parallel necks
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0.772.006 0.768.001 0.770.002
WITG0.772.006EA 418 EUR
WITG0.772.006 WITG0.768.001 WITG0.770.002
Flasks, round bottom, with three parallel necks
Ballons Flacons à fond rond
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Typical applications: distillation, extraction.