Imprimer
Flasks, pear shaped with standard ground joint
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0.664.100 0.662.250 0.664.050
WITG0.664.100EA 15.7 EUR
WITG0.664.100 WITG0.662.250 WITG0.664.050
Flasks, pear shaped with standard ground joint
Ballons Flacons piriformes
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
The conical geometry makes them ideal for small-scale reactions.