Imprimer
Flasks, round bottom, with two necks
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0.699.100 0.700.001 0.700.100
WITG0.699.100EA 34.2 EUR
WITG0.699.100 WITG0.700.001 WITG0.700.100
Flasks, round bottom, with two necks
Ballons Flacons à fond rond
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One vertical centre neck and one angled side neck (20°). With matt white labelling area.
Certifications: DIN 12394