Imprimer
Flasks, round bottom, with standard ground joints
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0.600.010 0.603.025 0.603.025.E 0.603.050.E 0.603.100.E 0.603.250.E 0.603.500.E 0.603.001.E 0.603.002.E 0.602.250.E 0.602.500.E 0.602.001.E 0.602.002.E 0.604.250.E 0.604.500.E 0.604.001.E 0.600.100.E 0.600.250.E 0.601.025.E 0.601.050.E 0.601.100.E 0.601.250.E 0.602.025.E 0.602.050.E 0.602.100.E 0.600.005.E 0.600.010.E 0.600.025.E 0.600.050.E
201-1371EA 99.5 EUR
201-1371 WITG0.603.025 WITG0.603.025.E WITG0.603.050.E WITG0.603.100.E WITG0.603.250.E WITG0.603.500.E WITG0.603.001.E WITG0.603.002.E WITG0.602.250.E WITG0.602.500.E WITG0.602.001.E WITG0.602.002.E WITG0.604.250.E WITG0.604.500.E WITG0.604.001.E WITG0.600.100.E WITG0.600.250.E WITG0.601.025.E WITG0.601.050.E WITG0.601.100.E WITG0.601.250.E WITG0.602.025.E WITG0.602.050.E WITG0.602.100.E WITG0.600.005.E WITG0.600.010.E WITG0.600.025.E WITG0.600.050.E
Flasks, round bottom, with standard ground joints
Ballons Flacons à fond rond
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck, useful for distillation and extraction.