Imprimer
Adapters with screw thread and standard gound joint cone
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0.550.001 0.550.010 0.550.040
WITG0.550.001EA 11.2 EUR
WITG0.550.001 WITG0.550.010 WITG0.550.040
Adapters with screw thread and standard gound joint cone
Adaptateurs Raccords à rodage conique
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Threaded adapter with DIN thread and standard ground joint cone.