Imprimer
Rotary evaporating flasks, with standard ground joints
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0.658.112 0.658.225
WITG0.658.112EA 128 EUR
WITG0.658.112 WITG0.658.225
Rotary evaporating flasks, with standard ground joints
Ballons Flacons pour évaporateurs rotatifs
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Pear-shaped, centered, with flat bottom.