Imprimer
Shaft guides, High Performance (HP), for standard taper ground glass joints
Fournisseur: Cowie
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
COWI005.0.0819.PKEA 232 EUR
COWI005.0.0819.PK COWI005.00.1029.PK COWI005.00.0824 COWI005.00.1024 COWI005.0.1945 COWI005.0.1229 COWI005.0.1029 COWI005.00.1029 COWI005.00.1645 COWIPC710.1925.PK COWI005.00.1945.PK COWI005.00.1229 COWI005.0.1024 COWIPC710.1025.PK COWI005.0.1045 COWIPC710.1625.PK COWIPC710.0825.PK COWI005.0.1029.PK COWI005.00.1045 COWI005.00.0819 COWI005.0.1945.PK COWIPC710.1019.PK COWI005.00.0619 COWI005.0.1024.PK COWI005.00.1945 COWI005.0.0819 COWI005.0.1034.PK COWI005.00.1645.PK COWIPC710.0819.PK COWI005.0.0619 COWIPC710.1225.PK COWI005.00.0824.PK COWI005.0.0824 COWI005.0.1229.PK COWI005.00.1045.PK COWI005.0.1634.PK COWI005.0.0624 COWI005.0.1634 COWI005.00.1634 COWI005.00.1024.PK COWI005.0.0824.PK COWI005.00.0624 COWI005.00.1229.PK COWI005.00.0819.PK COWI005.0.1034 COWI005.0.1045.PK COWI005.00.1634.PK
Shaft guides, High Performance (HP), for standard taper ground glass joints
Agitateurs Axes d'agitation
High Performance (HP) shaft guides provide an effective guide for use with glass and metal shaft stirrers over a wide range of temperatures without shedding particles from a specially formulated PTFE-PEEK seal whilst maintaining vacuum. The moulded-in PTFE-PEEK guide ring located at the end of the taper provides a guide to assist alignment of the shaft.
Shaft guides have an NPT thread for direct assembly into PTFE reactor lids or standard tapers for use with glass joints.
Attention: PEEK has reduced chemical resistance compared to PTFE and may be affected by acids, phenols and halogen based compounds.
Maximum recommended speeds are 500 rpm continuous or 800 rpm intermittent.