Imprimer
Pump tubing, Tygon®
Fournisseur: Heidolph Instruments GmbH & Co.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
525-60013-00 525-60014-00 525-60015-00 525-60016-00 525-60022-00 525-60023-00 525-60024-00 525-60025-00 525-60026-00 525-60027-00 525-60028-00 525-62000-00 525-63000-00 525-64000-00 525-65000-00 525-66000-00 525-69000-00 525-70027-00 525-70028-00 525-73000-00 525-74000-00 525-76000-00 525-80027-00 525-80028-00 525-83000-00 525-84000-00 525-85000-00 525-86000-00 525-89000-00 525-72000-00
HEID525-60013-00EA 26.8 EUR
HEID525-60013-00 HEID525-60014-00 HEID525-60015-00 HEID525-60016-00 HEID525-60022-00 HEID525-60023-00 HEID525-60024-00 HEID525-60025-00 HEID525-60026-00 HEID525-60027-00 HEID525-60028-00 224-1482 224-1478 224-1479 224-1480 224-1481 224-1483 HEID525-70027-00 HEID525-70028-00 228-5221 228-5222 228-5224 228-5226 228-5227 228-5229 228-5230 228-5231 228-5232 228-5233 HEID525-72000-00
Pump tubing, Tygon®
Tuyaux Transfer Tubing
Thermoplastic soft PVC, transparent.
  • Non-toxic, non-oxidizing
  • Good resistance to acids, bases and inorganic media
  • Very low gas permeability, good performance life
For standard applications.
Certifications: FDA and GLP conform.