Imprimer
Pump Tubing, Silicone
Fournisseur: Heidolph Instruments GmbH & Co.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
525-30014-00 525-30016-00 525-30024-00 525-30027-00 525-32000-00 525-33000-00 525-34000-00 525-35000-00 525-36000-00 525-39000-00
228-5206EA 71 EUR
228-5206 228-5208 228-5209 228-5212 228-5214 228-5215 228-5216 228-5217 228-5218 228-5219
Pump Tubing, Silicone
Tuyaux Transfer Tubing
Polydimethyl siloxane with siliceous earth and silicone additives; translucent white, excellent resistance to initial pressure.
  • Extremely smooth interior prevents bacterial growth
  • Biocompatible, minimal adsorption and absorption
  • Best flow properties, high temperature stability
  • Absolutely inert, softener-free
Platinum-coated silicone hose for use in pharmaceuticals and biology.