Imprimer
Glass density bottles, calibrated, supplied with DAkkS certifcate, BLAUBRAND®
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DAKKS43308 DAKKS43320 DAKKS43338
BRNDDAKKS43308EA 180 EUR
BRNDDAKKS43308 BRNDDAKKS43320 BRNDDAKKS43338
Glass density bottles, calibrated, supplied with DAkkS certifcate, BLAUBRAND®
Specific Gravity Bottles
Borosilicate glass 3,3, DIN ISO 3507, Gay-Lussac type.
  • Bottle and stopper (with capillary) are marked with unique matching identification number
  • Top of stopper ground and polished, for ease of cleaning
  • Individual certificate included
  • Reference temperature: 20 °C
The density bottles (pycnometers) are individually adjusted. The nominal capacity is indelibly engraved on each bottle. Each bottle is calibrated with its own stopper. The volume in cm3 is specified to a precision of 3 decimal places.