Imprimer
Ladies polo shirt, Acode® Basecamp
Fournisseur: FRISTADS KANSAS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1002213312XL 100221331L 100221331M 100221331S 100221331XL 1002219002XL 100221900L 100221900M 100221900S 100221900XL 1002219102XL 100221910L 100221910M 100221910S 100221910XL 1002219402XL 100221940L 100221940M 100221940S 100221940XL 100221930M
KWIN1002213312XLEA 20.3 EUR
KWIN1002213312XL KWIN100221331L KWIN100221331M KWIN100221331S KWIN100221331XL KWIN1002219002XL KWIN100221900L KWIN100221900M KWIN100221900S KWIN100221900XL KWIN1002219102XL KWIN100221910L KWIN100221910M KWIN100221910S KWIN100221910XL KWIN1002219402XL KWIN100221940L KWIN100221940M KWIN100221940S KWIN100221940XL KWIN100221930M
Ladies polo shirt, Acode® Basecamp
Tenue de travail
Heavy polo shirt made of 100% spunbounded and combed cotton piqué fabric. The fabric is pre-shrunk. The grey melange version is manufactured from 90% cotton and 10% viscose.
  • Fitted design
  • Four buttons
  • Rib-knit collar and cuffs
  • Side slits
Weight: 230 g/m²