Imprimer
Knee protectors, KS-955 and KT-957
Fournisseur: FRISTADS KANSAS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
100392940 100393930
KWIN100392940EA 42.8 EUR
KWIN100392940 KWIN100393930
Knee protectors, KS-955 and KT-957
Ergonomic Supports Knee Pads
For anyone who needs to work on their knees.
  • Can be combined with: all FRISTADS trousers that have 'knee pockets'
  • Not approved for commercial use
Length: 240 mm
Thickness: 13 or 25 mm
Width: 150 mm