Imprimer
Overalls, Fristads® PR54-820, design A, dark grey
Fournisseur: FRISTADS KANSAS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8529411312XL 852941131L 852941131M 852941131S 852941131XL 8529412102XL 852941210L 852941210M 852941210S 852941210XL 8529412312XL 852941231L 852941231M 852941231S 852941231XL 8529413312XL 852941331L 852941331M 852941331S 852941331XL 8529414402XL 852941440L 852941440M 852941440S 852941440XL 8529415202XL 852941520L 852941520M 852941520S 852941520XL 8529415302XL 852941530L 852941530M 852941530S 852941530XL 8529415392XL 852941539L 852941539M 852941539S 852941539XL 8529416402XL 852941640L 852941640M 852941640S 852941640XL 8529417302XL 852941730L 852941730M 852941730S 852941730XL 8529419402XL 852941940L 852941940M 852941940S 852941940XL 8529419412XL 852941941L 852941941M 852941941S 852941941XL
KWIN8529411312XLEA 0 EUR
KWIN8529411312XL KWIN852941131L KWIN852941131M KWIN852941131S KWIN852941131XL KWIN8529412102XL KWIN852941210L KWIN852941210M KWIN852941210S KWIN852941210XL KWIN8529412312XL KWIN852941231L KWIN852941231M KWIN852941231S KWIN852941231XL KWIN8529413312XL KWIN852941331L KWIN852941331M KWIN852941331S KWIN852941331XL KWIN8529414402XL KWIN852941440L KWIN852941440M KWIN852941440S KWIN852941440XL KWIN8529415202XL KWIN852941520L KWIN852941520M KWIN852941520S KWIN852941520XL KWIN8529415302XL KWIN852941530L KWIN852941530M KWIN852941530S KWIN852941530XL KWIN8529415392XL KWIN852941539L KWIN852941539M KWIN852941539S KWIN852941539XL KWIN8529416402XL KWIN852941640L KWIN852941640M KWIN852941640S KWIN852941640XL KWIN8529417302XL KWIN852941730L KWIN852941730M KWIN852941730S KWIN852941730XL KWIN8529419402XL KWIN852941940L KWIN852941940M KWIN852941940S KWIN852941940XL KWIN8529419412XL KWIN852941941L KWIN852941941M KWIN852941941S KWIN852941941XL
Overalls, Fristads® PR54-820, design A, dark grey
Tenue de travail
Work overalls made from 65% polyester and 35% cotton. With napped inner.
  • Zip fastening and plastic press studs
  • Two breast pockets with flap, two front pockets, two back pockets
  • Folding-rule pocket, leg pocket with flap, sleeve pocket, mobile phone pocket, pen pocket
  • Back pleats, adjustable elasticated waistband at the back, adjustable cuffs
  • Colour contrasting: Upper collar, inside front zip flap and pocket edging
Weight: 260 g/m²
Basic colour: Dark grey