Imprimer
Industrial jacket, Fristads® PR54-420, design A, dark grey
Fournisseur: FRISTADS KANSAS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8479411312XL 847941131L 847941131M 847941131S 847941131XL 847941131XS 8479412102XL 847941210L 847941210M 847941210S 847941210XL 847941210XS 8479412312XL 847941231L 847941231M 847941231S 847941231XS 8479413312XL 847941331L 847941331M 847941331S 847941331XL 847941331XS 8479414402XL 847941440XL 8479415202XL 847941520L 847941520M 847941520S 847941520XL 847941520XS 8479415302XL 847941530L 847941530M 847941530S 847941530XL 847941530XS 8479415392XL 847941539L 847941539M 847941539S 847941539XL 847941539XS 8479416402XL 847941640L 847941640M 847941640S 847941640XL 847941640XS 8479417302XL 847941730XL 8479419402XL 847941940L 847941940M 847941940S 847941940XL 847941940XS 8479419412XL 847941941L 847941941M 847941941S 847941941XL 847941941XS
113-5017EA 0 EUR
113-5017 113-03206 113-03207 113-03208 113-5016 113-03209 113-5013 113-03198 113-03199 113-03200 113-5012 113-03201 113-5019 113-03210 113-03211 113-03212 113-03213 113-5022 113-03214 113-03215 113-03216 113-5020 113-03217 113-5024 113-5023 113-5002 113-03178 113-03179 113-03180 113-5001 113-03181 113-5006 113-03186 113-03187 113-03188 113-5005 113-03189 113-5004 113-03182 113-03183 113-03184 113-5003 113-03185 113-5008 113-03190 113-03191 113-03192 113-5007 113-03193 113-5011 113-5009 113-5026 113-03222 113-03223 113-03224 113-5025 113-03225 113-5015 113-03202 113-03203 113-03204 113-5014 113-03205
Industrial jacket, Fristads® PR54-420, design A, dark grey
Tenue de travail
Thanks to its durability and high degree of comfort, the PR54 jacket is perfectly suited to workshops and industrial and service enterprises. The material is made from 65% polyester and 35% cotton. The inside is brushed and as a result conveys a pleasantly comfortably cotton feeling. The outside is also resistant to the roughest demands and will not fade even after countless washes.
  • Zip fastening and plastic press studs
  • Two breast pockets with flap, two front pockets
  • Sleeve pocket, mobile phone pocket, pen pocket, inner pocket
  • Back pleats, adjustable waist
  • 12 decorative colours available
Weight: 260 g/m²
Basic colour: Dark grey