Imprimer
Industrial jacket, Fristads® PR54-420, design A, navy blue
Fournisseur: FRISTADS KANSAS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8475391312XL 847539131L 847539131M 847539131S 847539131XL 847539131XS 8475392102XL 847539210L 847539210M 847539210S 847539210XL 847539210XS 8475392312XL 847539231L 847539231M 847539231S 847539231XL 847539231XS 8475393312XL 847539331L 847539331M 847539331S 847539331XL 847539331XS 8475394402XL 847539440L 847539440M 847539440S 847539440XL 847539440XS 8475395202XL 847539520L 847539520M 847539520S 847539520XL 847539520XS 8475395302XL 847539530L 847539530M 847539530S 847539530XL 847539530XS 8475395392XL 847539539L 847539539M 847539539S 847539539XL 847539539XS 8475396402XL 847539640L 847539640M 847539640S 847539640XL 847539640XS 8475397302XL 847539730L 847539730M 847539730S 847539730XL 847539730XS 8475399402XL 847539940L 847539940M 847539940S 847539940XL 847539940XS 8475399412XL 847539941L 847539941M 847539941S 847539941XL 847539941XS
113-03480EA 0 EUR
113-03480 113-03111 113-03112 113-03113 113-03479 113-03114 113-03476 113-03103 113-03104 113-03105 113-03475 113-03106 113-03482 113-03115 113-03116 113-03117 113-03481 113-03118 113-03484 113-03119 113-03120 113-03121 113-03483 113-03122 113-03486 113-03123 113-03124 113-03125 113-03485 113-03126 113-03466 113-03083 113-03084 113-03085 113-03465 113-03086 113-03470 113-03091 113-03092 113-03093 113-03469 113-03094 113-03468 113-03087 113-03088 113-03089 113-03467 113-03090 113-03472 113-03095 113-03096 113-03097 113-03471 113-03098 113-03474 113-03099 113-03100 113-03101 113-03473 113-03102 113-03488 113-03127 113-5393 113-03128 113-03487 113-03129 113-03478 113-03107 113-03108 113-03109 113-03477 113-03110
Industrial jacket, Fristads® PR54-420, design A, navy blue
Tenue de travail
Thanks to its durability and high degree of comfort, the PR54 jacket is perfectly suited to workshops and industrial and service enterprises. The material is made from 65% polyester and 35% cotton. The inside is brushed and as a result conveys a pleasantly comfortably cotton feeling. The outside is also resistant to the roughest demands and will not fade even after countless washes.
  • Zip fastening and plastic press studs
  • Two breast pockets with flap, two front pockets
  • Sleeve pocket, mobile phone pocket, pen pocket, inner pocket
  • Back pleats, adjustable waist
  • 12 decorative colours available
Weight: 260 g/m²
Basic colour: Marine