Imprimer
Shelving systems, standard, MetroMax Q®
Fournisseur: Metro International
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MQ2436G MQ2154G MQ1848G MQ2136G MQ2142G MQ2148G MQ1824G MQ1830G MQ1836G MQ1842G MQ2472G MQ2460G MQ2448G MQ2124G MQ2424G MQ2442G MQ1860G MQ1854G MQ2454G
METOMQ2436GEA 203 EUR
METOMQ2436G METOMQ2154G METOMQ1848G METOMQ2136G METOMQ2142G METOMQ2148G METOMQ1824G METOMQ1830G METOMQ1836G METOMQ1842G METOMQ2472G METOMQ2460G METOMQ2448G METOMQ2124G METOMQ2424G METOMQ2442G METOMQ1860G METOMQ1854G METOMQ2454G
Shelving systems, standard, MetroMax Q®
Mobilier Étagères
MetroMax Q® grid shelves feature removable polymer shelf mats with epoxy coated frames, a more durable and cleanable alternative to traditional epoxy coated wire shelving.
  • Quick to assemble
  • Quick to clean
Quick adjust shelves allow units to be easily reconfigured without tools on 25mm increments to maximize storage space. Vented mats allow air and light penetration and are easily removable for washing by hand or dish machines.
Informations sur les accessoires: A variety of optional accessories and casters will help contain, identify, organize, and transport supplies; contact your Avantor representative for more information.
Informations de commande: Each individual shelf, post, and solid mat overlay kit are sold separately. Mobile posts are fitted to accept casters (sold separately).