Imprimer
Glove box holders, Poxygrid®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F247390002 F247390003 F247390001
BELAF247390002EA 187 EUR
BELAF247390002 BELAF247390003 BELAF247390001
Glove box holders, Poxygrid®, SP Bel-Art
Distributeurs de gants et porte-boîtes à gants
Epoxy-coated, welded steel wire racks for one, two or three boxes of disposable gloves, wipes or tissues.
  • Glove box drops into an individual retaining compartment and lifts out easily for replacement
  • Designed for mounting to a vertical surface
  • Durable, blue epoxy coated racks are steam autoclavable at 121 °C