Imprimer
Dialysis device, Tube-A-Lyzer®, Spectra/Por®
Fournisseur: Spectrum Laboratories
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
137009 137042 137046 137050
734-1451EA 509 EUR
734-1451 734-1453 734-1455 734-1458
Dialysis device, Tube-A-Lyzer®, Spectra/Por®
Tuyaux Membranes de dialyse
Disposable and ready-to-use dynamic dialysis device.
  • Faster dialysis (4 to 12 hours)
  • Easy sample loading, testing and recovery
  • Prevents membrane polarisation and fouling
  • No heavy metals and sulphide contaminant
Device incorporates a semi-permeable membrane tubing that separates the sample chamber from the surrounding flow-through buffer chamber. The housing is designed to be a flow-through dialysate chamber that can be connected to a buffer source and a waste collection vessel or discharging directly to drain.