Imprimer
Closures for dialysis membrane tubing, universal
Fournisseur: Spectrum Laboratories
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
142153 142170 142172 142152 142150 142154 142155 142110 142250 142252 142112 142173 142113 142156 142174 142175 142176 142253
514-0647EA 245 EUR
514-0647 514-0651 514-0652 514-0646 514-0645 514-0648 514-0649 514-0642 514-0657 514-0658 514-0643 514-0653 514-0644 514-0650 514-0654 514-0655 514-0656 514-0659
Closures for dialysis membrane tubing, universal
Pinces Clips pour tubes
Polyamide, coloured
  • Available in a variety of colors for easy sample identification
  • For membrane types: Biotech CE, RC & PVDF; Standard RC
  • Sterilization: gamma irradiation or ethylene oxide (not autoclavable)
Specially designed with gentle sealing ridges, universal closures are the ideal all-purpose closure that creates a reliable seal with all membrane grades, types and thicknesses.