Imprimer
Buffer, TBS solution 10X concentrate (TRIS buffered saline) with phenol red, pH 7,4
Marques : Cayman Chemical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
600220-250 600220-500 600220-1
CAYM600220-250EA 71.4 EUR
CAYM600220-250 CAYM600220-500 CAYM600220-1
Buffer, TBS solution 10X concentrate (TRIS buffered saline) with phenol red, pH 7,4
Tampons Tampons d'électrophorèse Tampons de lavage
250 mM Tris-base, pH 7,4, 1,37 M sodium chloride, containing 27 mM KCl, 0,4 mM phenol red.
Suggested storage (long term) temperature: 4 °C.