Imprimer
Reagent bottles, with screw cap, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
844030 844029 844025 844026 844027
WHEA844030EA 67.9 EUR
WHEA844030 WHEA844029 WHEA844025 WHEA844026 WHEA844027
Reagent bottles, with screw cap, WHEATON®
Flacons Flacons en verre
Glass, amber, with blue screw cap.
Amber coloured to protect light sensitive substances.