Imprimer
Transport trolleys
Fournisseur: BOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
11418 11417
BOCH11418EA 1710 EUR
BOCH11418 BOCH11417
Transport trolleys
Chariots
These transport trolleys are made of stainless steel with two guide and two fixed rolls.
  • Surface matt finish
  • Robust pushing handle
Load capacity: 300 kg