Imprimer
Tubing connectors, '5 in 1', SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F196550000 F196570000
BELAF196550000EA 133 EUR
BELAF196550000 BELAF196570000
Tubing connectors, '5 in 1', SP Bel-Art
Raccords et connecteurs
PP
  • Fits 5 sizes of tubing and will connect the same size tubing on both ends, or will act as a reducer for any combination within the range
  • Inside diameter increases with notch size when unneeded length is cut off for unrestricted flow