Imprimer
Ampoules, standard, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
176759 176762
WHEA176759EA 432 EUR
WHEA176759 WHEA176762
Ampoules, standard, WHEATON®
Ampoules
Ampoules are made from low potassium borosilicate glassare.
  • Consistent with low background counts
Commonly used for packaging certified standards.
Certifications: USP Type I, ASTM E 438 Type I, Class A