Imprimer
Electrophoresis gel plate racks, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F135950000 F135960000
581-7611EA 363 EUR
581-7611 BELAF135960000
Electrophoresis gel plate racks, SP Bel-Art
Portoirs Portoirs pour plaques d'électrophorèse
Versatile solution for drying and storing gel plates. Rugged, A-shaped racks that securely supports glass gel plates in an upright position for rapid drying and easy access.
  • 10 plate capacity