Imprimer
Cavity slides
Fournisseur: MARIENFELD
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1320000 1320202 1320002 1320102
631-9475EA 31.2 EUR
631-9475 630-1613 630-1611 630-1612
Cavity slides
Slides Cavity Slides
Soda-lime glass of hydrolytic class 3 cavity slides.
Carefully ground and polished for in vitro diagnostic (IVD) applications.
Certifications: IVD 98/79/EC