Hexylphosphonic acid
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Attention
|
CAS No.: 4721-24-8
Molecular Formula: C6H13P(O)(OH)2
Formula Weight: 166.16
Physical Form: Powder
Appearance: White to off-white
Melting Point: 108-110°
MDL No.: MFCD00013961
Formule:
C₆H₁₅O₃P Point de fusion: 108…110 °C Température de stockage: Température ambiante |
Numéro MDL:
MFCD00013961 Numéro CAS: 4721-24-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...