Imprimer

Glycylglycine (Gly-Gly)

Fournisseur: Molekula
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

MOLEM49774934EA 0 EUR
MOLEM49774934
Glycylglycine (Gly-Gly)
Protéines et Peptides
  • EINECS: 209-127-8
  • GHS symbol and signal word: GHS07 and warning
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR