Imprimer
Sleeves for ground joints, with gripping ring, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H933-03 H933-05 H933-06 H933-09 H933-10 H933-11 H933-12 H933-13 H933-14 H933-15 H933-04 H933-01 H933-07 H933-02
BOHLH933-03EA 11.6 EUR
BOHLH933-03 BOHLH933-05 BOHLH933-06 BOHLH933-09 BOHLH933-10 BOHLH933-11 BOHLH933-12 BOHLH933-13 BOHLH933-14 BOHLH933-15 BOHLH933-04 BOHLH933-01 BOHLH933-07 BOHLH933-02
Sleeves for ground joints, with gripping ring, BOLA
Manchons et raccords pour joints NS Manchettes pour rodage de raccordement
Reusable sleevesmade of PTFE, with gripping ring and sealing rings on the outside.
  • Chemical resistant and temperature resistant from −200 to +250°C
  • High-vacuum tight sealing
  • Low leak rate: <0,0001 mbar× L/s
  • Solid construction and sealing rings are easy accessible