Imprimer
Sleeves for ground joints, with ribs, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H930-01 H930-02 H930-03 H930-04 H930-05 H930-06 H930-07 H930-08 H930-09 H930-10 H930-11 H930-13 H930-14 H930-15 H932-10 H932-07 H932-06 H932-05 H932-03
BOHLH930-01EA 6.7 EUR
BOHLH930-01 BOHLH930-02 BOHLH930-03 201-6125 201-6126 BOHLH930-06 201-6127 BOHLH930-08 BOHLH930-09 201-6129 BOHLH930-11 BOHLH930-13 201-6130 BOHLH930-15 BOHLH932-10 BOHLH932-07 BOHLH932-06 BOHLH932-05 BOHLH932-03
Sleeves for ground joints, with ribs, BOLA
Manchons et raccords pour joints NS Manchettes pour rodage de raccordement
PTFE.
  • Universal chemical resistance
  • Suitable vacuum
  • Temperature resistant from −200 to +250 °C
With sealing rings ('ribs') on the outside. Without gripping ring and therefore usable with joint clamps. For vacuum up to 0,1 mbar, solid construction for continued use and sealing rings are easily accessible and therefore easy to clean.