Imprimer
Mesh woven filters, stainless steel, Spectra Mesh®
Fournisseur: Spectrum Laboratories
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
145825 145934 145827 145936 146439 148985
SPEC145825EA 369 EUR
SPEC145825 SPEC145934 SPEC145827 SPEC145936 SPEC146439 SPEC148985
Mesh woven filters, stainless steel, Spectra Mesh®
Filtres Filtres à membrane
These woven filters are constructed of individual strands woven into a mesh screen, characterised by precise mesh openings, percent open area and mesh thickness. Available in five material types with different physical, chemical and thermal characteristics to select from depending on your application. Available in discs or square sheets.
Water affinity: Hydrophilic
Strength/durability: Very high
Adsorption: High
Best resistance: Corrosives
pH resistance: 4 to 12
Thermal stability: up to 180 ºC
Sterilisation: Autoclavable / irradiation