Imprimer
Funnels with short stem, heavy duty, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F146970000 F146960000
BELAF146970000EA 128 EUR
BELAF146970000 BELAF146960000
Funnels with short stem, heavy duty, SP Bel-Art
Entonnoirs Entonnoirs pour la filtration Entonnoirs filtrants
PP, translucent.
  • With ridges to help prevent vapor lock
  • Durable and versatile
  • Autoclavable at 121 °C
The heavy duty funnel is ideal for continuous laboratory work.