Imprimer
Magnetic sink strainer, Spinbar®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F377870000 F377870001
442-0592EA 82.8 EUR
442-0592 442-0593
Magnetic sink strainer, Spinbar®, SP Bel-Art
Agitateurs Barreaux magnétiques
Spinbar® Magnetic Sink Strainer is a simple solution to prevent your Spinbar® Magnetic Stirring Bars from going down the sink drain.
  • Four strong magnets attract and hold stir bars
  • Four legs on the underside prevent strainer from being dislodged by flowing liquid
  • Long slits in the sides and skirt allow fluids to drain rapidly
  • Wide skirt allows use in a variety of sink drains
  • Straight edge on one side allows it to be placed near the sink wall
  • Flexible and chemical-resistant
Available in blue or green.