Imprimer
Chairs, Fabric
Fournisseur: Kango
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6NN35GHLP00705 6NN35GDLV02705 6NN35GDLV00705 6NN35GBLP00705 6NN35GHLP02705 6NN35GBLR00705 6NN35GHLR02705 6NN35NBPR00705 6NN35NBPP00705 6NN35GTLP01705 6NN40GBLR00705 6NN40GBLP00705 6NN40GHLP00705 6NN40GHLP01705 6NN35NHPP00705 6NN35NHPP02705
132-7894EA 365 EUR
132-7894 132-8095 132-8096 132-8434 132-8435 132-8437 BIEN6NN35GHLR02705 BIEN6NN35NBPR00705 BIEN6NN35NBPP00705 BIEN6NN35GTLP01705 BIEN6NN40GBLR00705 BIEN6NN40GBLP00705 BIEN6NN40GHLP00705 BIEN6NN40GHLP01705 BIEN6NN35NHPP00705 BIEN6NN35NHPP02705
Chairs, Fabric
Chaises
Chairs with high density comfortable foam and fabric coating
  • Ergonomically designed seat
  • Available with castors or glides
  • Can also be equipped with a footring
  • Gas lift column with height adjustment and shok absorber
  • 5-branch scuff resistant reinforced base
Chairs:
Shaped backrest with rounded edges
Articulated backrest, adjustable in height and depth (adjustable chairs)Independently adjustable seat and backrest tilt with one fingertip lever thanks to an asynchronous mechanism (for asynchronous chairs)Rectangular seat
Dimensions of the seat (W×D): 435×390 mm
Dimensions of the backrest (W×H): 410×250 mm