Imprimer
Sample spoons, for balances, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F367070000 F367060000
BELAF367070000EA 0 EUR
BELAF367070000 BELAF367060000
Sample spoons, for balances, SP Bel-Art
Cuillères
Stainless steel, with or without PTFE coating. These trough-style balance spoons enable aasily transfer small quantities of materials.
  • Available in plain, polished stainless steel or with PTFE coating
  • Square trough-type spoon end: 44×8,9 mm