Imprimer
Measuring cylinders, tall form, with handle, Holdfast®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F284611000 F284612000
612-4970EA 0 EUR
612-4970 612-4971
Measuring cylinders, tall form, with handle, Holdfast®, SP Bel-Art
Éprouvettes Éprouvettes graduées
PP, translucent, tall form, with handle.
  • Provides a secure, solid grip for safer pouring
  • Easy-to-read double scale with raised graduations
  • Hexagonal base prevents tipping and rolling
  • Will not create a meniscus