Imprimer
Tanks, rectangular, heavy duty, with top flanges, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H340850000 H340940000 H340950000 H340930000 H340920000
BELAH340850000EA 735 EUR
BELAH340850000 BELAH340940000 BELAH340950000 BELAH340930000 BELAH340920000
Tanks, rectangular, heavy duty, with top flanges, SP Bel-Art
Récipients Récipients rectangulaires
These seamless, rotational-molded PE tanks are lightweight and easy-to-handle.
  • Available with or without a faucet
  • Acid and alkali resistant
  • With wide top flanges, include covers
Informations de commande: Can be supplied with spigots, fittings, dams, partitions etc. Contact local Avantor sales office for quotations on special sizes and fabrication in PP and PVC.