Imprimer
Safety spectacles, uvex meteor
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9134.005 9134.002
UVEX9134.005EA 37.7 EUR
UVEX9134.005 UVEX9134.002
Safety spectacles, uvex meteor
Safety Glasses
The dual lens glasses with close fitting frame provide reliable protection and protection against particles from below.
  • Anatomically shaped nose bridge
  • Side shields and upper eye rim protection integrated in frame design
  • Side arms individually adjustable in four different lengths
  • Adjustable comfortable soft-form ear pieces
Certifications: EN 166
Informations de livraison: Supplied with case and microfibre cloth.