Imprimer
Rack for cuvettes, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F185160787 F185160000
634-8291EA 96.3 EUR
634-8291 BELAF185160000
Rack for cuvettes, SP Bel-Art
Portoirs Portoirs pour cuvettes
Racks, made of PP, translucent. Hold fragile spectralphotometer cuvettes in complete safety.
  • Only the corners of the cuvettes touch the racks
  • Accommodate 12 cuvettes of 10 mm light path
  • Will not scratch quartz cuvettes
  • F18516-0787 features Stack Rack™ interlocking end plates for secure stacking
These two tier racks support cuvettes with a minimum of physical contact, greatly reducing the chance of damage.