Imprimer
ESD safety shoes, lace-up, uvex motion style, 6998
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6998.8/43 6998.8/36 6998.8/37 6998.8/38 6998.8/39 6998.8/40 6998.8/41 6998.8/42 6998.8/44 6998.8/45 6998.8/46 6998.8/47 6998.8/48 6989.8/36 6989.8/37 6989.8/38 6989.8/42 6989.8/43 6989.8/39 6989.8/40 6989.8/41 6989.8/44 6989.8/45 6989.8/46 6989.8/47 6989.8/48 6989.8/49 6989.8/50
UVEX6998.8/43EA 150 EUR
UVEX6998.8/43 UVEX6998.8/36 UVEX6998.8/37 UVEX6998.8/38 UVEX6998.8/39 UVEX6998.8/40 UVEX6998.8/41 UVEX6998.8/42 UVEX6998.8/44 UVEX6998.8/45 UVEX6998.8/46 UVEX6998.8/47 UVEX6998.8/48 114-5535 114-5536 114-5537 114-5541 114-5542 114-5538 114-5539 114-5540 114-5543 114-5544 114-5545 114-5546 114-5547 114-5548 114-5549
ESD safety shoes, lace-up, uvex motion style, 6998
Chaussures et bottes
Ultralight, perforated ESD safety shoe made from breathable high-tech microvelour, soft padded collar and tongue, breathable distance mesh and an extremely flexible dual density PUR sole (slip-, abrasion- and cut-resistant, resistant to oil and petrol and withstands brief exposure to heat of approx. 120 °C). Free of lacquer wetting inhibitors such as silicones or phthalates.
  • Climazone with highly breathable lining and outer materials
  • Toecap made from steel (200 J)
  • Anti-twist heel cap for better stability and protection from ankle twisting
  • 3D hydroflex® foam insole for exceptional shock absorption in the heel and under the front of the foot
  • Comfortable removable insole, anti-static, with a moisture control system
  • Extremely flexible, light weight PUR outsole, which is abrasion-, cut-, and slip-resistant
  • Fulfils ESD requirements: resistance to ground <35 megaohms
Suitable for people allergic to chrome.
Colour: Black/grey or Black/red.
Certifications: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S1, SRC