Imprimer
Sterilising trays and covers, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H162640000 H162630000 H162620000 H162600000 H162610000
BELAH162640000EA 180 EUR
BELAH162640000 BELAH162630000 BELAH162620000 BELAH162600000 BELAH162610000
Sterilising trays and covers, SP Bel-Art
Plateaux/Récipients/Pots Bacs de stérilisation
These lightweight and corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.
  • Ideal for sterilising instruments, bottles containing agar, deionised water, saline and other reagents, and for safely cleaning glassware in sinks
  • Polypropylene covers are 3,2 mm thick with a 25,4 mm skirt that fits snugly over the rim of the tray
  • Trays and lids are steam autoclavable at 121 ºC