Imprimer
Portoirs pour incubateur, Stak-A-Tray™
Fournisseur: Bel-Art
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Stak-a-Tray™
BELAH186101440EA 308 EUR
BELAH186101440 631-9778 631-9777 631-9775
Portoirs pour incubateur, Stak-A-Tray™
Portoirs Portoirs pour incubateurs
These are stainless steel modular rack and tray system that maximizes storage for petri dishes and cell culture flasks in incubators, cold rooms or on the bench.
  • Perforated trays allow air flow and slide easily in the rack
  • Stack up to five racks for 20 levels of storage
  • Each tier accommodates one large or two small trays
All parts are stainless steel and autoclavable at 121 ºC (250 ºF).
Informations de commande: Racks and trays are sold separately.