Imprimer
Pipeline filters, with filter disc, VitraPOR®
Fournisseur: ROBU GLASFILTER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
23601 23602 23603 23604 23605 23900 23901 23902 23903 23904 23905 23305 23304 23600 23301 23302 23303 23300
ROBU23601EA 231 EUR
ROBU23601 ROBU23602 ROBU23603 ROBU23604 ROBU23605 ROBU23900 ROBU23901 ROBU23902 ROBU23903 ROBU23904 ROBU23905 ROBU23305 ROBU23304 ROBU23600 ROBU23301 ROBU23302 ROBU23303 ROBU23300
Pipeline filters, with filter disc, VitraPOR®
Filters Filter Tubes
These filters are used for in-line filtration of gas lines to remove solid impurities (e.g. dust). Pipeline filters are available with filter discs in diameters from 30 to 90 mm.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
  • With tube adapters for connection
Made of borosilicate glass 3.3.