Imprimer
Filtering tiles, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N1610-10 N1616-30 N1616-20 N1616-10
BOHLN1610-10EA 849 EUR
BOHLN1610-10 BOHLN1616-30 BOHLN1616-20 BOHLN1616-10
Filtering tiles, BOLA
Filtres Carrés filtrants
These filtering tiles are made of microporous PTFE.
  • Suitable for cutting or stamping
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C