Imprimer

Eusolex® OS

Marques : Merck
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

1.06949.9025 1.06949.0100 1.06949.1000
1.06949.9025EA 0 EUR
1.06949.9025 1.06949.0100 1.06949.1000
Eusolex® OS
Eusolex®

Eusolex® OS is a clear, liquid UV-B filter, oil soluble with excellent dissolving properties for solid UV filters.


INCI name: Ethylhexyl Salicylate

Point d'ébullition: 189…190 °C (28 hPa)
Densité: 1.013 g/cm3 (25 °C)
Point éclair: 141 °C
Température de stockage: Température ambiante
EINECS: 204-263-4

Commander maintenant

Résultats des tests de spécification

Color colorless to yellowish
Appearance clear liquid
Odour weak odour
Assay Octisalate (GC) 95.0 - 105.0 %
Chromatography purity conforms every single impurity <= 0,5 A
% sum of impurities <= 2,0 A%
Methyl salicylate (GC) ≤ 100 ppm
Identity (retention time) conforms
Identity (IR-spectrum) conforms
Refractive index (n 20/D) 1.500 - 1.503
Density (d 25/25) 1.011 - 1.016
Specific absorption (305 nm; 1 %; 1 cm; methanol) 173 - 182
Heavy metals (as Pb) ≤ 0.001 %
As (Arsenic) ≤ 0.0005 %
Residual solvent Methanol (HS-GC) ≤ 2000
Acidity ≤ 1,0 ml
Saponification value 215 - 230
Microbial Purity (Total Viable Aerobic Count) ≤ 10² CFU/g
E.coli absent in 1 g
Pseudomonas aeruginosa absent in 1 g
Staphylococcus aureus absent in 1 g
Candida albicans absent in 1 g
Salmonella species absent in 10 g
We confirm that the product meets the purity requirements defined in USP and JSQI by applying our internal testing methods which might deviate from those under USP and JSQI.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR