Imprimer
Laboratory glass cleaner, deconex® 22 PF
Fournisseur: Borer Chemie
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
501500.00-S10P 501500.00-D15P
148-5235EA 174 EUR
148-5235 DECO501500.00-D15P
Laboratory glass cleaner, deconex® 22 PF
Produits nettoyants Détergents et Nettoyants de Surfaces
Alkaline cleaning powder for automated cleaning of laboratory glassware und reusable laboratory material.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • With a broad cleaning spectrum and excellent results even at low concentrations
  • High cleaning performance due to excellent dispersing capabilities
  • Phosphate-free, suited for phosphate-critical applications
  • EDTA-, NTA-, chlorine- and surfactant-free, environmentally friendly, no release of phosphates and surfactants into waste water
  • Rinsing with an acidic deconex® cleaner is recommended as the completion of the cleaning process